tert-Butyl methyl(pyrrolidin-3-yl)carbamate

Lieferant: Apollo Scientific

Synonyme: 3-(N-Boc-N-methylamino)pyrrolidine, 3-(N-tert-Butoxycarbonyl-N-methylamino)pyrrolidine

OR451220-1G
APOSOR451220-1GEA 91.5 EUR
APOSOR451220-1G
tert-Butyl methyl(pyrrolidin-3-yl)carbamate
tert-Butyl methyl(pyrrolidin-3-yl)carbamate
Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00191665
CAS-Nummer: 172478-00-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR