4-Methyltetrahydro-2H-pyran-4-ol
Lieferant: Apollo Scientific
OR451221-5G
APOSOR451221-5GEA
130
EUR
APOSOR451221-5G
4-Methyltetrahydro-2H-pyran-4-ol
4-Methyltetrahydro-2H-pyran-4-ol
Formel:
C₆H₁₂O₂ Molecular Weight: 116,1600 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13193576 CAS-Nummer: 7525-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...