4-(Aminomethyl)tetrahydro-2H-pyran-4-carboxylic acid hydrochloride

Lieferant: Apollo Scientific

OR451224-1G
APOSOR451224-1GEA 873 EUR
APOSOR451224-1G
4-(Aminomethyl)tetrahydro-2H-pyran-4-carboxylic acid hydrochloride
4-(Aminomethyl)tetrahydro-2H-pyran-4-carboxylic acid hydrochloride
Formel: C₇H₁₃NO₃·ClH
Molecular Weight: 195,644 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25371776
CAS-Nummer: 1485427-12-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR