4-[(Boc-amino)methyl]tetrahydro-2H-pyran-4-carboxylic acid

Lieferant: Apollo Scientific

Synonyme: 4-(((tert-Butoxycarbonyl)amino)methyl)tetrahydro-2H-pyran-4-carboxylic acid

OR451225-1G
APOSOR451225-1GEA 1190 EUR
APOSOR451225-1G
4-[(Boc-amino)methyl]tetrahydro-2H-pyran-4-carboxylic acid
4-[(Boc-amino)methyl]tetrahydro-2H-pyran-4-carboxylic acid
Formel: C₁₂H₂₁NO₅
Molecular Weight: 259,299 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09264199
CAS-Nummer: 946761-11-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR