1-Aminoindan-1-carboxylic acid
Lieferant: Apollo Scientific
OR451226-1G
APOSOR451226-1GEA
1020
EUR
APOSOR451226-1G
1-Aminoindan-1-carboxylic acid
1-Aminoindan-1-carboxylic acid
Formel:
C₁₀H₁₁NO₂ Molecular Weight: 177,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00800595 CAS-Nummer: 3927-71-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...