Spiro[imidazolidine-4,1'-indane]-2,5-dione

Lieferant: Apollo Scientific

OR451227-5G
APOSOR451227-5GEA 800 EUR
APOSOR451227-5G
Spiro[imidazolidine-4,1'-indane]-2,5-dione
Spiro[imidazolidine-4,1'-indane]-2,5-dione
Formel: C₁₁H₁₀N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03951077
CAS-Nummer: 6252-98-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR