Boc-1-aminoindan-1-carboxylic acid

Lieferant: Apollo Scientific

Synonyme: 1-(Boc-amino)-1-indanecarboxylic Acid

OR451228-1G
APOSOR451228-1GEA 188 EUR
APOSOR451228-1G
Boc-1-aminoindan-1-carboxylic acid
Boc-1-aminoindan-1-carboxylic acid
Formel: C₁₅H₁₉NO₄
Molecular Weight: 277,32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00800596
CAS-Nummer: 214139-26-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR