(S)-2-(1H-Imidazol-1-ylmethyl)-morpholine trihydrochloride

Lieferant: Apollo Scientific

OR451232-1G
APOSOR451232-1GEA 1540 EUR
APOSOR451232-1G
(S)-2-(1H-Imidazol-1-ylmethyl)-morpholine trihydrochloride
(S)-2-(1H-Imidazol-1-ylmethyl)-morpholine trihydrochloride
Formel: C₈H₁₃N₃O·3ClH
Molecular Weight: 276,591 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28975220
CAS-Nummer: 278789-01-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR