(R)-4-Boc-2-(aminomethyl)morpholine
Lieferant: Apollo Scientific
Synonyme:
(R)-tert-Butyl 2-(aminomethyl)morpholine-4-carboxylate
OR451233-500MG
APOSOR451233-500MGEA
844
EUR
APOSOR451233-500MG
(R)-4-Boc-2-(aminomethyl)morpholine
(R)-4-Boc-2-(aminomethyl)morpholine
Formel:
C₁₀H₂₀N₂O₃ Molecular Weight: 216,277 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13189480 CAS-Nummer: 1174913-80-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...