4-(3,5-Dimethyl-1H-pyrazol-4-yl)-benzoic acid

Lieferant: Apollo Scientific

OR451237-1G
APOSOR451237-1GEA 0 EUR
APOSOR451237-1G
4-(3,5-Dimethyl-1H-pyrazol-4-yl)-benzoic acid
4-(3,5-Dimethyl-1H-pyrazol-4-yl)-benzoic acid
Formel: C₁₂H₁₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03425153
CAS-Nummer: 185252-69-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR