(R)-1-Boc-2-ethylpiperazin
Lieferant: Apollo Scientific
OR451238-5G
APOSOR451238-5GEA
342
EUR
APOSOR451238-5G
(R)-1-Boc-2-ethylpiperazin
(R)-1-Boc-2-ethylpiperazin
Formel:
C₁₁H₂₂N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03411920 CAS-Nummer: 393781-70-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...