a-(4-Chlorophenyl)-4-Boc-1-piperazineacetic acid
Lieferant: Apollo Scientific
OR451239-1G
APOSOR451239-1GEA
960
EUR
APOSOR451239-1G
a-(4-Chlorophenyl)-4-Boc-1-piperazineacetic acid
a-(4-Chlorophenyl)-4-Boc-1-piperazineacetic acid
Formel:
C₁₇H₂₃ClN₂O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659337 CAS-Nummer: 885273-01-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...