3-Bromo-4-nitropyridine

Lieferant: Apollo Scientific
Achtung

OR45124-25G
APOSOR45124-25GEA 104 EUR
APOSOR45124-25G
3-Bromo-4-nitropyridine
3-Bromo-4-nitropyridine
Formel: C5H3BrN2O2
Molecular Weight: 202.99 g/mol
Schmelzpunkt: 66…67 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03085767
CAS-Nummer: 89364-04-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR