1-Boc-4-(2-amino-1-(4-bromophenyl)ethyl)piperazine

Lieferant: Apollo Scientific

OR451240-1G
APOSOR451240-1GEA 960 EUR
APOSOR451240-1G
1-Boc-4-(2-amino-1-(4-bromophenyl)ethyl)piperazine
1-Boc-4-(2-amino-1-(4-bromophenyl)ethyl)piperazine
Formel: C₁₇H₂₆BrN₃O₂
Molecular Weight: 384,311 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10015110
CAS-Nummer: 1018544-60-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR