2-(N-Boc-piperazino)-2-(4-methoxyphenyl)ethylamine

Lieferant: Apollo Scientific

OR451241-1G
APOSOR451241-1GEA 960 EUR
APOSOR451241-1G
2-(N-Boc-piperazino)-2-(4-methoxyphenyl)ethylamine
2-(N-Boc-piperazino)-2-(4-methoxyphenyl)ethylamine
Formel: C₁₈H₂₉N₃O₃
Molecular Weight: 335,441 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10015092
CAS-Nummer: 444892-61-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR