2-(N-Boc-piperazino)-2-(4-chlorophenyl)ethylamine
Lieferant: Apollo Scientific
OR451242-1G
APOSOR451242-1GEA
960
EUR
APOSOR451242-1G
2-(N-Boc-piperazino)-2-(4-chlorophenyl)ethylamine
2-(N-Boc-piperazino)-2-(4-chlorophenyl)ethylamine
Formel:
C₁₇H₂₆ClN₃O₂ Molecular Weight: 339,86 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10015095 CAS-Nummer: 444892-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...