7,8-Dihydro-5H-pyrano[4,3-d]pyrimidin-2-amine

Lieferant: Apollo Scientific

OR451245-1G
APOSOR451245-1GEA 531 EUR
APOSOR451245-1G
7,8-Dihydro-5H-pyrano[4,3-d]pyrimidin-2-amine
7,8-Dihydro-5H-pyrano[4,3-d]pyrimidin-2-amine
Formel: C₇H₉N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD15202267
CAS-Nummer: 1211486-21-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR