2,3,4,5-Tetrahydropyrazolo[1,5-b][1,2]oxazepine-7-carboxylic acid

Lieferant: Apollo Scientific

OR451247-250MG
APOSOR451247-250MGEA 388 EUR
APOSOR451247-250MG
2,3,4,5-Tetrahydropyrazolo[1,5-b][1,2]oxazepine-7-carboxylic acid
2,3,4,5-Tetrahydropyrazolo[1,5-b][1,2]oxazepine-7-carboxylic acid
Formel: C₈H₁₀N₂O₃
Molecular Weight: 182,177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28975211
CAS-Nummer: 1934470-69-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR