3,4-Dibromtoluol 98%
Lieferant: Apollo Scientific
Synonyme:
1,2-Dibromo-4-methylbenzene
OR4513-100G
APOSOR4513-100GEA
87
EUR
APOSOR4513-100G
3,4-Dibromtoluol 98%
3,4-Dibromtoluol
Formel:
C₇H₆Br₂ |
MDL:
MFCD00079744 CAS-Nummer: 60956-23-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...