1-(Cyanmethyl)pyridiniumchlorid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR451315-100G
OR451315-25G
OR451315-5G
APOSOR451315-100GEA
0
EUR
APOSOR451315-100G
APOSOR451315-25G
APOSOR451315-5G
1-(Cyanmethyl)pyridiniumchlorid 97%
1-(Cyanomethyl)pyridinium chloride
Formel:
C₇H₇ClN₂ Schmelzpunkt: 169…174 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00031933 CAS-Nummer: 17281-59-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...