(2S)-2-Methylmorpholine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR45132-1G
APOSOR45132-1GEA 66.7 EUR
APOSOR45132-1G
(2S)-2-Methylmorpholine hydrochloride
(2S)-2-Methylmorpholine hydrochloride
Formel: C₅H₁₁NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD11506701
CAS-Nummer: 1147108-99-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR