2-(5-Chloro-1H-benzimidazol-2-yl)ethanamine dihydrochloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR451401-1G
OR451401-5G
APOSOR451401-1GEA
102
EUR
APOSOR451401-1G
APOSOR451401-5G
2-(5-Chloro-1H-benzimidazol-2-yl)ethanamine dihydrochloride
2-(5-Chloro-1H-benzimidazol-2-yl)ethanamine dihydrochloride
Formel:
C₉H₁₀ClN₃·2ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD03419439 CAS-Nummer: 91003-86-0 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...