Ethyl 3-azabicyclo[3.1.0]hexane-6-carboxylate hydrochloride 95+%

Lieferant: Apollo Scientific
Achtung

OR45142-250MG
APOSOR45142-250MGEA 188 EUR
APOSOR45142-250MG
Ethyl 3-azabicyclo[3.1.0]hexane-6-carboxylate hydrochloride 95+%
Ethyl 3-azabicyclo[3.1.0]hexane-6-carboxylate hydrochloride
Formel: C₈H₁₃NO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD11111227
CAS-Nummer: 1211510-15-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR