4-Iodo-1-methyl-1H-pyrazol-3-amine

Lieferant: Apollo Scientific
Gefahr

OR451431-1G OR451431-25G OR451431-5G
APOSOR451431-1GEA 267 EUR
APOSOR451431-1G APOSOR451431-25G APOSOR451431-5G
4-Iodo-1-methyl-1H-pyrazol-3-amine
4-Iodo-1-methyl-1H-pyrazol-3-amine
Formel: C₄H₆IN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00847646
CAS-Nummer: 150187-16-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR