4-(2-Bromoethyl)benzonitrile

Lieferant: Apollo Scientific
Achtung

OR45144-5G
APOSOR45144-5GEA 79.9 EUR
APOSOR45144-5G
4-(2-Bromoethyl)benzonitrile
4-(2-Bromoethyl)benzonitrile
Formel: C₉H₈BrN
Lagertemperatur: Kühlschrank
MDL: MFCD12025077
CAS-Nummer: 72054-56-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR