4-Bromo-1H-pyrazol-3-amine hydrobromide

Lieferant: Apollo Scientific
Achtung

OR451440-10G OR451440-1G OR451440-5G
APOSOR451440-10GEA 179 EUR
APOSOR451440-10G APOSOR451440-1G APOSOR451440-5G
4-Bromo-1H-pyrazol-3-amine hydrobromide
4-Bromo-1H-pyrazol-3-amine hydrobromide
Formel: C₃H₄BrN₃·BrH
Molecular Weight: 242.9 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08456823
CAS-Nummer: 16461-99-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR