Methyl-4-(chloromethyl)-5-methylisoxazole-3-carboxylate

Lieferant: Apollo Scientific
Achtung

OR451447-1G OR451447-5G OR451447-10G
APOSOR451447-1GEA 212 EUR
APOSOR451447-1G APOSOR451447-5G APOSOR451447-10G
Methyl-4-(chloromethyl)-5-methylisoxazole-3-carboxylate
Methyl-4-(chloromethyl)-5-methylisoxazole-3-carboxylate
Formel: C₇H₈ClNO₃
Molecular Weight: 189.6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14702898
CAS-Nummer: 1177292-31-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR