3-Bromo-4-chloro-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

OR451453-1G OR451453-5G
APOSOR451453-1GEA 78.3 EUR
APOSOR451453-1G APOSOR451453-5G
3-Bromo-4-chloro-1H-pyrazole
3-Bromo-4-chloro-1H-pyrazole
Formel: C₃H₂BrClN₂
Lagertemperatur: Kühlschrank
MDL: MFCD30079725
CAS-Nummer: 27258-12-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR