2-Bromo-5-ethoxy-4-hydroxybenzaldehyde

Lieferant: Apollo Scientific
Achtung

OR451460-1G
APOSOR451460-1GEA 69.6 EUR
APOSOR451460-1G
2-Bromo-5-ethoxy-4-hydroxybenzaldehyde
2-Bromo-5-ethoxy-4-hydroxybenzaldehyde
Formel: C₉H₉BrO₃
Molecular Weight: 245.08 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01119132
CAS-Nummer: 340216-58-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR