Benzo[d]thiazole-2-carbonitrile 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,3-Benzothiazole-2-carbonitrile

OR45147-1G OR45147-250MG
APOSOR45147-1GEA 270 EUR
APOSOR45147-1G APOSOR45147-250MG
Benzo[d]thiazole-2-carbonitrile 98%
Benzo[d]thiazole-2-carbonitrile
Formel: C₈H₄N₂S
Molecular Weight: 160.196 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08848199
CAS-Nummer: 2602-85-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR