2,6-Dichloro-3-iodopyridin-4-amine ≥97%

Lieferant: Apollo Scientific

OR45155-250MG
APOSOR45155-250MGEA 82.8 EUR
APOSOR45155-250MG
2,6-Dichloro-3-iodopyridin-4-amine ≥97%
2,6-Dichloro-3-iodopyridin-4-amine
Formel: C₅H₃Cl₂IN₂
Molecular Weight: 288,901 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD29918898
CAS-Nummer: 1235873-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR