2-Cyclopropylbenzenesulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR45172-1G
OR45172-250MG
OR45172-5G
APOSOR45172-1GEA
0
EUR
APOSOR45172-1G
APOSOR45172-250MG
APOSOR45172-5G
2-Cyclopropylbenzenesulphonyl chloride
2-Cyclopropylbenzenesulphonyl chloride
Formel:
C₉H₉ClO₂S Lagertemperatur: Kühlschrank |
MDL:
MFCD31666885 CAS-Nummer: 1772586-39-6 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...