5-Bromo-2-phenyl-1H-indole
Lieferant: Apollo Scientific
|
Achtung
|
OR45176-1G
APOSOR45176-1GEA
74.1
EUR
APOSOR45176-1G
5-Bromo-2-phenyl-1H-indole
5-Bromo-2-phenyl-1H-indole
Formel:
C₁₄H₁₀BrN Molecular Weight: 272.14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04151729 CAS-Nummer: 83515-06-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...