(2-Brom-4,5-dimethoxyphenyl)essigsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-4,5-dimethoxyphenylacetic acid
OR4518-5G
APOSOR4518-5GEA
231
EUR
APOSOR4518-5G
(2-Brom-4,5-dimethoxyphenyl)essigsäure
(2-Brom-4,5-dimethoxyphenyl)essigsäure
Formel:
C₁₀H₁₁BrO₄ Molecular Weight: 275.1 g/mol Schmelzpunkt: 94…96 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016819 CAS-Nummer: 4697-62-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...