2-Bromo-5-iodo-3-methylbenzyl alcohol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Bromo-5-iodo-3-methylphenyl)methanol

OR45195-250MG OR45195-1G OR45195-5G
APOSOR45195-250MGEA 190 EUR
APOSOR45195-250MG APOSOR45195-1G APOSOR45195-5G
2-Bromo-5-iodo-3-methylbenzyl alcohol 95%
2-Bromo-5-iodo-3-methylbenzyl alcohol
Formel: C₈H₈BrIO
Molecular Weight: 326.96 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD32868532
CAS-Nummer: 2091676-26-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR