Boc-D-Tyr(ME)-OH 97%
Lieferant: Apollo Scientific
Synonyme:
Boc-D-Phe(4-OMe)-OH, N-Boc-4-methoxy-D-phenylalanine, N-(tert-Butoxycarbonyl)-4-methoxy-D-phenylalanine
OR452000-25G
OR452000-5G
APOSOR452000-25GEA
336
EUR
APOSOR452000-25G
APOSOR452000-5G
Boc-D-Tyr(ME)-OH 97%
Boc-D-Tyr(ME)-OH
Formel:
C₁₅H₂₁NO₅ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00065604 CAS-Nummer: 68856-96-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...