Boc-D-Tyr(ME)-OH 97%

Lieferant: Apollo Scientific

Synonyme: Boc-D-Phe(4-OMe)-OH, N-Boc-4-methoxy-D-phenylalanine, N-(tert-Butoxycarbonyl)-4-methoxy-D-phenylalanine

OR452000-25G OR452000-5G
APOSOR452000-25GEA 336 EUR
APOSOR452000-25G APOSOR452000-5G
Boc-D-Tyr(ME)-OH 97%
Boc-D-Tyr(ME)-OH
Formel: C₁₅H₂₁NO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD00065604
CAS-Nummer: 68856-96-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR