(R)-2-(3-Chlorophenyl)glycine 97%

Lieferant: Apollo Scientific

Synonyme: (R)-2-Amino-2-(3-chlorophenyl)acetic acid, H-D-Phg(3-Cl)-OH

OR452013-1G
APOSOR452013-1GEA 289 EUR
APOSOR452013-1G
(R)-2-(3-Chlorophenyl)glycine 97%
(R)-2-(3-Chlorophenyl)glycine
Formel: C₈H₈ClNO₂
Molecular Weight: 185.608 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07371715
CAS-Nummer: 25698-37-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR