7-Methyl-L-tryptophan 97%

Lieferant: Apollo Scientific

Synonyme: (S)-2-Amino-3-(7-methyl-1H-indol-3-yl)propanoic acid, H-Trp(7-Me)-OH

OR452024-250MG
APOSOR452024-250MGEA 149 EUR
APOSOR452024-250MG
7-Methyl-L-tryptophan 97%
7-Methyl-L-tryptophan
Formel: C₁₂H₁₄N₂O₂
Molecular Weight: 218.252 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06797706
CAS-Nummer: 33468-36-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR