4-Chloro-L-tryptophan 97%
Lieferant: Apollo Scientific
OR452030-1G
OR452030-250MG
APOSOR452030-1GEA
0
EUR
APOSOR452030-1G
APOSOR452030-250MG
4-Chloro-L-tryptophan 97%
4-Chloro-L-tryptophan
Formel:
C₁₁H₁₁ClN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797630 CAS-Nummer: 52448-14-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...