4-Chloro-D-tryptophan 97%

Lieferant: Apollo Scientific

OR452031-1G OR452031-250MG OR452031-500MG
APOSOR452031-1GEA 0 EUR
APOSOR452031-1G APOSOR452031-250MG APOSOR452031-500MG
4-Chloro-D-tryptophan 97%
4-Chloro-D-tryptophan
Formel: C₁₁H₁₁ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10565751
CAS-Nummer: 27542-41-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR