3-(Boc-amino)-3-(2-methylphenyl)propionsäure 97%

Lieferant: Apollo Scientific

Synonyme: 3-((tert-Butoxycarbonyl)amino)-3-(o-tolyl)propanoic acid

OR452039-1G
APOSOR452039-1GEA 916 EUR
APOSOR452039-1G
3-(Boc-amino)-3-(2-methylphenyl)propionsäure 97%
3-(Boc-amino)-3-(2-methylphenyl)propionsäure
Formel: C₁₅H₂₁NO₄
Molecular Weight: 279,3315 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02090696
CAS-Nummer: 284493-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR