Boc-DL-β-leucine 97%

Lieferant: Apollo Scientific

Synonyme: 3-(Boc-amino)-4-methylpentanoic acid, DL-3-((tert-Butoxycarbonyl)amino)-4-methylpentanoic acid, Boc-DL-β-HoVal-OH

OR452041-1G
APOSOR452041-1GEA 78.3 EUR
APOSOR452041-1G
Boc-DL-β-leucine 97%
Boc-DL-beta-leucine
Formel: C₁₁H₂₁NO₄
Molecular Weight: 231.29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02258127
CAS-Nummer: 248924-39-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR