N-Fmoc-3-hydroxy-L-phenylalanine 97%

Lieferant: Apollo Scientific

Synonyme: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-hydroxyphenyl)propanoic acid

OR452043-1G OR452043-250MG OR452043-500MG
APOSOR452043-1GEA 654 EUR
APOSOR452043-1G APOSOR452043-250MG APOSOR452043-500MG
N-Fmoc-3-hydroxy-L-phenylalanine 97%
N-Fmoc-3-hydroxy-L-phenylalanine
Formel: C₂₄H₂₁NO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD07371961
CAS-Nummer: 178432-48-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR