N-Fmoc-3-hydroxy-L-phenylalanine 97%
Lieferant: Apollo Scientific
Synonyme:
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-hydroxyphenyl)propanoic acid
OR452043-1G
OR452043-250MG
OR452043-500MG
APOSOR452043-1GEA
654
EUR
APOSOR452043-1G
APOSOR452043-250MG
APOSOR452043-500MG
N-Fmoc-3-hydroxy-L-phenylalanine 97%
N-Fmoc-3-hydroxy-L-phenylalanine
Formel:
C₂₄H₂₁NO₅ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07371961 CAS-Nummer: 178432-48-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...