4-[1-(Boc-amino)ethyl]benzoic acid 97%

Lieferant: Apollo Scientific

Synonyme: 4-(1-((tert-Butoxycarbonyl)amino)ethyl)benzoic acid

OR452050-1G
APOSOR452050-1GEA 130 EUR
APOSOR452050-1G
4-[1-(Boc-amino)ethyl]benzoic acid 97%
4-[1-(Boc-amino)ethyl]benzoic acid
Formel: C₁₄H₁₉NO₄
Molecular Weight: 265.305 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505982
CAS-Nummer: 895577-21-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR