2-(3-Chlorophenoxy)-2-methylpropanoic acid 97%

Lieferant: Apollo Scientific

OR452060-1G
APOSOR452060-1GEA 393 EUR
APOSOR452060-1G
2-(3-Chlorophenoxy)-2-methylpropanoic acid 97%
2-(3-Chlorophenoxy)-2-methylpropanoic acid
Formel: C₁₀H₁₁ClO₃
Molecular Weight: 214,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01695660
CAS-Nummer: 17413-73-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR