2-(2-Chlorophenoxy)-benzoic acid 97%

Lieferant: Apollo Scientific

OR452062-1G OR452062-250MG
APOSOR452062-1GEA 568 EUR
APOSOR452062-1G APOSOR452062-250MG
2-(2-Chlorophenoxy)-benzoic acid 97%
2-(2-Chlorophenoxy)-benzoic acid
Formel: C₁₃H₉ClO₃
Molecular Weight: 248.662 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06260646
CAS-Nummer: 36809-08-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR