2-(2-Chlorophenoxy)-benzoic acid 97%
Lieferant: Apollo Scientific
OR452062-1G
OR452062-250MG
APOSOR452062-1GEA
568
EUR
APOSOR452062-1G
APOSOR452062-250MG
2-(2-Chlorophenoxy)-benzoic acid 97%
2-(2-Chlorophenoxy)-benzoic acid
Formel:
C₁₃H₉ClO₃ Molecular Weight: 248.662 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06260646 CAS-Nummer: 36809-08-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...