L-Alanine cyclobutylmethyl ester hydrochloride 97%

Lieferant: Apollo Scientific

OR452064-1G OR452064-5G
APOSOR452064-1GEA 340 EUR
APOSOR452064-1G APOSOR452064-5G
L-Alanine cyclobutylmethyl ester hydrochloride 97%
L-Alanine cyclobutylmethyl ester hydrochloride
Formel: C₈H₁₅NO₂·ClH
Molecular Weight: 193.671 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27952567
CAS-Nummer: 261909-76-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR